Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Caspase-9 Substrate I, Fluorogenic
A fluorogenic substrate for caspase-9 (ICE-LAP6, Mch6).
$164.00 - $450.00
Specifications
Color | White |
---|---|
Formulation | Supplied as a trifluoroacetate salt. |
Description | 95% by HPLC |
Target Proteins | A fluorogenic substrate for caspase-9 |
Molecular Formula | C33H38F3N7O11 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
21-876-51MG
|
MilliporeSigma™
2187651MG |
1 mg |
Each for $164.00
|
|
|||||
21-876-55MG
|
MilliporeSigma™
2187655MG |
5 mg |
Each for $450.00
|
|
|||||
Description
Peptide Sequence: Ac-Leu-Glu-His-Asp-AFCSpecifications
White | |
95% by HPLC | |
C33H38F3N7O11 | |
DMSO | |
Solid |
Supplied as a trifluoroacetate salt. | |
A fluorogenic substrate for caspase-9 | |
765.7g/mol | |
Protect from Light, Protect from Moisture |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title