Learn More
MilliporeSigma™ Calbiochem™ Adenosine A1 Receptor Antagonist II, PSB-36
A highly potent and selective antagonist of adenosine A1 receptor (Ki = 124 and 700 pM for rat and human A1R, respectively).
Manufacturer:  MilliporeSigma™ 11914810MG
Catalog No. 11-914-810MG
Add to cart
Description
Description
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
‘Lysine-20’
Specifications
Specifications
≥98% by HPLC
2-8°C, Protect from Light
524944-72-7
Tan powder
10mg
C21H30N4O3