Learn More
MilliporeSigma™ Calbiochem™ Adenosine A2A/A1 Receptor Antagonist
The Adenosine A2A/A1 Receptor Antagonist controls the biological activity of Adenosine A2A/A1 Receptor.
Manufacturer:  MilliporeSigma™ 11676510MG
Catalog No. 11-676-510MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
97% by HPLC
Hygroscopic
Protect from Light
C23H22N4O3·2HCl
DMSO
Light yellow solid
10mg
475.4g/mol