Learn More
MilliporeSigma™ Calbiochem™ Adenosine A2A Receptor Antagonist II, SCH-58261
A highly potent and selective antagonist of adenosine A2A receptor (Ki = 6
Manufacturer:  MilliporeSigma™ 11915010MG
Catalog No. 11-915-010MG
Add to cart
Description
Description
A highly potent and selective antagonist of adenosine A2A receptor (Ki = 600 pM). Does not affect the activity of other adenosine receptors in any significant manner (Ki = 287nM; 5.0µM, and > 10µM for A1, A2B, and A3 receptors, respectively). Displays neuroprotective effects in cerebral ischaemia in rats, and shown to counteract Parkinsonian-like muscle rigidity in rats.
Specifications
Specifications
99% by HPLC
Protect from Light
A2A Receptor
DMSO
Off-white solid
10mg
C18H15N7O