Learn More
MilliporeSigma™ Calbiochem™ Adenosine A2B Receptor Antagonist I, MRS 1754
A p-cyanoanilide xanthine derivative that acts as a highly selective, high affinity antagonist of adenosine A2B receptor (A2B R;
Manufacturer:  MilliporeSigma™ 11915110MG
Catalog No. 11-915-110MG
Add to cart
Description
Description
A p-cyanoanilide xanthine derivative that acts as a highly selective, high affinity antagonist of adenosine A2B receptor (A2BR; Ki = 1.97nM for human A2BR). Does not significantly affect other adenosine receptors (Ki = 403 nM, 503 nM, and 570nM for human A1R, A2AR, and A3R, respectively). Exhibits some antagonist action on rat A1R (Ki = 16.8nM). Shown to abolish the cardioprotective effects of NECA (Cat. No. 119140).
Specifications
Specifications
99% by HPLC
Protect from Light
Adenosine A2B Receptor
486.5g/mol
White solid
10mg
C26H26N6O4
DMSO