Learn More
MilliporeSigma™ Calbiochem™ Adenosine Kinase Inhibitor
The Adenosine Kinase Inhibitor, also referenced under CAS 214697-26-4, controls the biological activity of Adenosine Kinase.
Manufacturer:  MilliporeSigma™ 1168905MG
Catalog No. 11-689-05MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
90% by HPLC
Hygroscopic
Protect from Light
Adenosine Kinase
C22H19BrN6O·2HCl
DMSO
Yellow solid
5mg
214697-26-4
536.3g/mol