Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine Triphosphate, 2-Methylthio-, Tetrasodium Salt

An ATP analog that induces aortic smooth muscle relaxation in rat (EC50 = 240 nM).

Manufacturer:  MilliporeSigma™ 1191265MG

Catalog No. 11-912-65MG


Add to cart

Description

Description

An ATP analog that induces aortic smooth muscle relaxation in rat (EC50 = 240 nM).
Specifications

Specifications

90% by HPLC
Toxicity: Standard Handling

Store at ≤ -70°C


Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C.
43170-89-4
641.2g/mol
White solid
5mg
C11H14N5O13P3SNa4·xH2O
H2O
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit