Learn More
MilliporeSigma™ Calbiochem™ Caspase-3 Substrate IV, Fluorogenic
Fluorogenic caspase-3 substrate.
Manufacturer:  MilliporeSigma™ 2641505MG
 View more versions of this product
Catalog No. 26-415-05MG
Add to cart
Description
Description
Peptide Sequence: Z-Asp-Glu-Val-Asp-AFC
Specifications
Specifications
95% by HPLC
5mg
C36H38F3N5O14
DMSO
White or off-white solid
Fluorogenic caspase-3 substrate.
821.7g/mol