Learn More
MilliporeSigma™ Calbiochem™ Caspase-3 Substrate VII, Fluorogenic
Fluorogenic caspase-3 substrate.
Supplier:  MilliporeSigma™ 2641515MG
 View more versions of this product
Catalog No. 26-415-15MG
Add to Cart
Description
Description
Peptide Sequence: Ac-Asp-Glu-Val-Asp-AFC
Specifications
Specifications
5 mg
Protect from Light, Protect from Moisture
95% by HPLC
C30H34F3N5O13
DMSO
Lyophilized Solid
White
Fluorogenic caspase-3 substrate
729.6g/mol