Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenylyl-imidodiphosphate, Tetralithium Salt Available on GSA/VA Contract for Federal Government customers only.

A non-hydrolyzable ATP analog that acts as a competitive inhibitor of ATP-dependent enzyme systems.

Manufacturer:  MilliporeSigma™ 12000225MG

Catalog No. 12-000-225MG

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Contaminants: Lithium: ≤4.5%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

>85% by UV
Hygroscopic
ATP-sensitive Ca2+-activated K+ channels.
C10H13N6O12P3·4Li
H2O
White solid
25mg
72957-42-7
529.9g/mol
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit