Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ (±)-Bay K 8644
Synthetic dihydropyridine derivative that acts as an active Ca2+ slow channel agonist in neuroendocrine, muscle, thyroid and other cell types.
$113.00 - $356.00
Specifications
Color | Yellow |
---|---|
Description | 98% by HPLC |
Target Proteins | L-type Ca2+ channel |
CAS | 93468-89-4 |
Molecular Formula | C16H15F3N2O4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-687-81MG
|
MilliporeSigma™
1968781MG |
1 mg |
Each for $113.00
|
|
|||||
19-687-85MG
|
MilliporeSigma™
1968785MG |
5 mg |
Each for $356.00
|
|
|||||
Description
Synthetic dihydropyridine derivative that acts as an active Ca2+ slow channel agonist in neuroendocrine, muscle, thyroid and other cell types. Prolongs single channel open time without affecting the close time. An L-type Ca2+ channel agonist. Composed of two optical isomers. The (-)-enantiomer has strong vasoconstrictive, positive inotropic, and Ca2+ agonistic properties, whereas the (+)-enantiomer has weakly vasodilating, negative inotropic, and Ca2+ antagonistic properties. The net effect of the racemic mix is that of the negative enantiomer. Promotes β-cell proliferation/regeneration.Specifications
Yellow | |
L-type Ca2+ channel | |
C16H15F3N2O4 | |
EtOH or MeOH | |
Solid |
98% by HPLC | |
93468-89-4 | |
356.3g/mol | |
Protect from light |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title