EMD Millipore Calbiochem™ AEBSF, Hydrochloride Available on GSA/VA Contract for Federal Government customers only.

AEBSF, CAS 30827-99-7, is a specific irreversible inhibitor of chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. A nontoxic alternative to PMSF and DFP.

Manufacturer: EMD Millipore 101500100MG

 View more versions of this product

Catalog No. 10-150-0100MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Moisture
    Target Proteins chymotrypsin
    CAS 30827-99-7
    Molecular Formula C8H10FNO2S·HCl
    Formula Weight 239.7g/mol
    Solubility H2O
    Description >97% by HPLC
    Format White to off-white solid
    Quantity 100mg

    Packaged Under Inert Gas

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit