Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Agarose, Type C
Purified non-ionic fraction of agar
Supplier: MilliporeSigma™ 121852100GM
Description
- Forms gel with mechanical qualities at very low concentrations
- Can be mounted on microscope slides or transparent plastic
- When dried, films are elastic, durable
- Show little or no deterioration on staining with either protein or lipoprotein dyes
- Gelling temperature: 40 to 43°C
- Contaminants Sulfate: ≤0.3%
- CAS: 9012-36-6
Specifications
Agarose | |
100 g | |
−15° to 30°C |
Safety and Handling
ShelfLife : Stock solutions is Stable for up to 1 month at 20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title