Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Monastrol
A potent, cell-permeable, non-tubulin-interacting mitosis inhibitor.
$123.19 - $404.57
Specifications
Color | White |
---|---|
Target Proteins | Mitotic kinesin Eg5 |
CAS | 254753-54-3 |
Molecular Formula | C14H16N2O3S |
Formula Weight | 292.4g/mol |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
47-587-91MG
|
MilliporeSigma™
4758791MG |
1 mg |
Each for $123.19
|
|
|||||
47-587-95MG
|
MilliporeSigma™
4758795MG |
5 mg |
Each for $404.57
|
|
|||||
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
White | |
254753-54-3 | |
292.4g/mol | |
Protect from light | |
95% by HPLC |
Mitotic kinesin Eg5 | |
C14H16N2O3S | |
DMSO | |
Solid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title