Learn More
MilliporeSigma™ Calbiochem™ Mitochondrial Pyruvate Carrier Inhibitor, UK5099
A cell-permeable α-cyanocinnamate compound that acts as a rapid, active site thiol modifying, time-dependent and non-competitive inhibitor of bi-directional pyruvate transport across plasma membranes in yeast, bacteria, Drosophila, and humans (IC50 = 50nM in rat heart mitochondria; Ki ≤ 1µM) with excellent selectivity over monocarboxylates and
Manufacturer:  MilliporeSigma™ 5.04817.0001
 View more versions of this product
Catalog No. 50-481-70001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A cell-permeable α-cyanocinnamate compound that acts as a rapid, active site thiol modifying, time-dependent and non-competitive inhibitor of bi-directional pyruvate transport across plasma membranes in yeast, bacteria, Drosophila, and humans (IC50 = 50nM in rat heart mitochondria; Ki ≤ 1µM) with excellent selectivity over monocarboxylates and anions. Shown to significantly decrease pyruvate-driven respiration, enhance glucose uptake and activate AMPK (2µM in human myocytes and skeletal muscle myotubes).
Specifications
Specifications
99% by HPLC
Protect from Light
mitochondria
DMSO
Bright yellow solid
5mg
C18H12N2O2