Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ p-Nitroblue Tetrazolium Chloride
NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50 = 3-4 μM).
$91.00 - $259.00
Specifications
Color | Yellow |
---|---|
Description | 98% by TLC |
Target Proteins | NOS |
CAS | 298-83-9 |
Molecular Formula | C40H30Cl2N10O6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
48-423-51GM
|
MilliporeSigma™
4842351GM |
1 g |
Each for $259.00
|
|
|||||
48-423-5250MG
|
MilliporeSigma™
484235250MG |
250 mg |
Each for $91.00
|
|
|||||
Description
NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50 = 3-4μM). A well-known scavenger of superoxide anions. Useful as a substrate for alkaline phosphatase, often in conjunction with BCIP (Cat. No. 203788).Specifications
Yellow | |
NOS | |
C40H30Cl2N10O6 | |
MeOH or H2O | |
Crystalline Solid |
98% by TLC | |
298-83-9 | |
817.6g/mol | |
Hygroscopic, Protect from light |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title