Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Pifithrin-α
A cell-permeable chemical inhibitor of p53.
$120.00 - $213.00
Specifications
Color | Off-white |
---|---|
Description | 95% by HPLC |
Target Proteins | PS3 |
CAS | 63208-82-2 |
Molecular Formula | C16H18N2OS·HBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-613-25MG
|
MilliporeSigma™
5061325MG |
5 mg |
Each for $120.00
|
|
|||||
50-613-210MG
|
MilliporeSigma™
50613210MG |
10 mg |
Each for $213.00
|
|
|||||
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Off-white | |
PS3 | |
C16H18N2OS·HBr | |
DMSO | |
Solid |
95% by HPLC | |
63208-82-2 | |
367.3g/mol | |
Protect from light |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title