EMD Millipore™ Calbiochem™ Albumin, Human Serum, Fraction V Available on GSA/VA Contract for Federal Government customers only.

Prepared under non-denaturing conditions by modification of Cohn procedure

Manufacturer: EMD Millipore™ 1266810GM

 View more versions of this product

Catalog No. 12-668-10GM

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Disclaimers:

For Research Use Only


    Specifications

    Format Solid
    Source Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
    Quantity 10g
    Storage Requirements 2° to 8°C
    Purity ≥95% by SDS PAGE

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit