Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PARP Inhibitor VIII, PJ34
The PARP Inhibitor VIII, PJ34, also referenced under CAS 344458-15-7, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.
$120.00 - $321.00
Specifications
Color | White |
---|---|
Description | 98% by HPLC |
Target Proteins | PARP |
CAS | 344458-15-7 |
Molecular Formula | C17H17N3O2·HCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
52-815-05MG
|
MilliporeSigma™
5281505MG |
5 mg |
Each for $321.00
|
|
|||||
52-815-01MG
|
MilliporeSigma™
5281501MG |
1 mg |
Each for $120.00
|
|
|||||
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
White | |
PARP | |
C17H17N3O2·HCl | |
H2O | |
Solid |
98% by HPLC | |
344458-15-7 | |
331.8g/mol | |
Hygroscopic, Protect from light |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title