EMD Millipore™ Calbiochem™ Ampicillin, Sodium Salt Available on GSA/VA Contract for Federal Government customers only.

Suitable for use in research that uses ampicillin-resistant plasmids

Manufacturer: EMD Millipore™ 1712545GM

 View more versions of this product

Catalog No. 17-125-45GM

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Product Type Ampicillin
    Quantity 5g
    Storage Requirements 2- 8°C
    Format White solid

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit