Learn More
MilliporeSigma™ Calbiochem™ APETx2, Anthopleura elegantissima, Recombinant, E. coli
Originally isolated from sea anemone Anthopleura elegantissima , the 42 amino-acid toxin APETx2 is shown to block acidification-induced activation of homotrimeric acid-sensing ion channel composed of rat or human ASIC3, but not rat ASIC1a, ASIC1b, or ASIC2a, in a reversible man
Supplier: MilliporeSigma™ 5.00527.0001
Description
Originally isolated from sea anemone Anthopleura elegantissima , the 42 amino-acid toxin APETx2 is shown to block acidification-induced activation of homotrimeric acid-sensing ion channel composed of rat or human ASIC3, but not rat ASIC1a, ASIC1b, or ASIC2a, in a reversible manner. APETx2 is also shown to inhibit the Na+-dependent, alkalization-induced nonconventional channel activity seen in human, but not rat, ASIC3. APETx2 in vivo efficacy is demonstrated in various rat pain induction models (100μL 2.2μM APETx2 via i.m. injection). Reported to display much reduced potency against Kv3.4 and little or no effect toward hERG, Kv2.2, Kv3.1, Kv4.2, or Kv4.3 channel activity.Specifications
Supplied as a trifluoroacetate salt. | |
ASIC3 | |
4561g/mol | |
Protect from light | |
100 μg |
95% by HPLC | |
C196H280N54O61S6 | |
Aqueous buffer | |
Lyophilized |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.