EMD Millipore Calbiochem™ Apicidin, Fusarium sp.

A potent, cell-permeable inhibitor of histone deacetylase (IC50= 700 pM for parasitic histone deactetylase) that also exibits antiprotozoal and potential anti-malarial properties.

Manufacturer: EMD Millipore 1782765MG

 View more versions of this product

Catalog No. 17-827-65MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins Histone deacetylase
    Molecular Formula C34H49N5O6
    Formula Weight 623.8g/mol
    Solubility EtOH and DMSO
    Description 95% by HPLC
    Format Off-white solid
    Quantity 5mg

    • Packaged Under Inert Gas
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit