Learn More
EMD Millipore Calbiochem™ Aprotinin, Bovine Lung, Solution Available on GSA/VA Contract for Federal Government customers only.
Aprotinin, Bovine Lung, Solution, CAS 9087-70-1, is a competitive, reversible, heat and acid-stable inhibitor of proteolytic and esterolytic activities.
Manufacturer:  EMD Millipore 616399500KU
 View more versions of this product
Catalog No. 61-639-9500KU
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
6,500 KIU/mL
Liquid
chymotrypsin
6512g/mol
In isotonic NaCl, aseptically filled.
500 ku
9087-70-1