Learn More
MilliporeSigma™ Calbiochem™ ARE Activator, BTZO-1
The ARE Activator, BTZO-1 modulates the biological activity of ARE. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  MilliporeSigma™ 18131510MG
Catalog No. 18-131-510MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
98% by HPLC
Protect from Light
C13H8N2OS
DMSO
Off-white solid
10mg
240.3g/mol