Learn More
MilliporeSigma™ Calbiochem™ Aromatase Inhibitor II, Letrozole
The Aromatase Inhibitor II, Letrozole, also referenced under CAS 112809-51-5, controls the biological activity of Aromatase.
Manufacturer:  MilliporeSigma™ 18254125MG
Catalog No. 18-254-125MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
99% by HPLC
Protect from Light
112809-51-5
285.3g/mol
White powder
25mg
C17H11N5
DMSO