Learn More
MilliporeSigma™ Calbiochem™ Ascomycin, Streptomyces hygroscopicus
An ethyl analog of FK506 with similar immunosuppressant properties.
Manufacturer:  MilliporeSigma™ 1828001MG
Catalog No. 18-280-01MG
Add to cart
Description
Description
An ethyl analog of FK506 with similar immunosuppressant properties. Acts by binding to immunophilins. Inhibits the synthesis of Th-1 and Th-2 type cytokines in target cells.
Specifications
Specifications
90% by HPLC
1mg
11011-38-4
792g/mol
White crystalline solid
Immunophilins
C43H69NO12
DMSO or EtOH