Learn More
MilliporeSigma™ Calbiochem™ ATP Assay Kit
Detects ATP production in various enzymatic reactions, detects low level bacterial contamination in specific samples
Supplier: MilliporeSigma™ 1191071KIT
Includes: One vial of Luciferase ATP Monitoring Enzyme, Enzyme Reconstitution buffer, 1 bottle of Nucleotide Releasing Reagent, one vial of ATP standard and user protocol
Description
- Luminescence method of detection
- Applications: ATP quantified in blood, milk, urine, soil, sludge
- Fully automatic for high throughput (10s/sample)
- Extremely sensitive
- Detects 10 to 100 mammalian cells/well
- Packaging: Glass Bottle
Specifications
ATP Assay Kit | |
One vial of Luciferase ATP Monitoring Enzyme, Enzyme Reconstitution Buffer, 1 bottle of Nucleotide Releasing Reagent, one vial of ATP Standard, and user protocol | |
1 Kit |