Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ ATR Inhibitor III, ETP-46464
A cell-permeable quinoline-containing heterotricyclic compound that acts as a potent inhibitor against mTOR, ATR, DNA-PK, PI 3-Kα, and ATM (IC50
Supplier: MilliporeSigma™ 5.00508.0001
Description
A cell-permeable quinoline-containing heterotricyclic compound that acts as a potent inhibitor against mTOR, ATR, DNA-PK, PI 3-Kα, and ATM (IC50 = 0.6, 14, 36, 170, and 545 nM, respectively) and preferentially suppresses radiation-induced cellular ATR activity (>90% at 500nM) over ATM or DNA-PK activity (IC50 > 5μM) in U2OS cells, while exhibiting much reduced or little potency toward a panel of 26 other kinases (>55% inhibition at 5μM). Shown to synergize with UCN-01 (Cat. No. 539644) in replicative stress induction in S-phase replicating U2OS (1.5-, 61-, and 190-fold of control RS, respectively, with 1μM ETP-46464, 50nM UCN-01, or combined treatment).Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Specifications
White | |
ATR | |
DMSO | |
Powder |
99% by HPLC | |
C30H22N4O2 | |
Protect from light | |
10 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title