EMD Millipore Calbiochem™ Bafilomycin A1, Streptomyces griseus Available on GSA/VA Contract for Federal Government customers only.

Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.

Manufacturer: EMD Millipore 1960001SET

 View more versions of this product

Catalog No. 19-600-01SET

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Storage Requirements Protect from Light
    Target Proteins vacuolar type H+ ATPase
    Molecular Formula C35H58O9
    Formula Weight 622.8g/mol
    Solubility DMSO or EtOH
    Description 90% by HPLC
    Format White solid
    Quantity 1 set
    CAS 88899-55-2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit