Learn More
EMD Millipore Calbiochem™ Bafilomycin A1, Streptomyces griseus Available on GSA/VA Contract for Federal Government customers only.
Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.
Manufacturer:  EMD Millipore 19600010UG
 View more versions of this product
Catalog No. 19-600-010UG
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
90% by HPLC
Protect from Light
vacuolar type H+ ATPase
C35H58O9
DMSO or EtOH
White solid
10μg
88899-55-2
622.8g/mol