Learn More
MilliporeSigma™ Calbiochem™ Bcl-2 Binding Peptide, Cell-Permeable
A cell-permeable peptide that binds to Bcl-2 with high affinity (IC50 = 130 nM in a competition binding study) and induces apoptosis.
Manufacturer:  MilliporeSigma™ 1972201MG
Catalog No. 19-722-01MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Peptide Sequence: CH3(CH2)8CONH-Lys-Asn-Leu-Trp-Ala-Ala-Gln-Arg-Tyr-Gly-Arg-Glu-Leu-Arg-Arg-Met-Ser-Asp-Glu-Phe-Glu-Gly-Ser-Phe-Lys-Gly-Leu-OH
  • Cell Permeable
Specifications
Specifications
Supplied as a trifluoroacetate salt.
Off-white solid
1mg
C153H240N44O42S
DMSO
95% by HPLC
Hygroscopic
Bcl-2
3399.9g/mol