Learn More
MilliporeSigma™ Calbiochem™ Bcr-abl Inhibitor
The Bcr-abl Inhibitor, also referenced under CAS 778270-11-4, controls the biological activity of Bcr-abl. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  MilliporeSigma™ 1972215MG
Catalog No. 19-722-15MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
97% by HPLC
Protect from Light
Bcr-abl-expressing cells in Ba/F3.p210
C18H13F3N4O2
DMSO or EtOH
White solid
5mg
778270-11-4
374.3g/mol