Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Bikinin
A cell-permeable nonsteroidal pyridinylamido compound that acts as an ATP-competitive inhibitor.
Supplier: MilliporeSigma™ 20098025MG
Description
Packaged Under Inert GasSpecifications
25 mg | |
Solid | |
90% by HPLC | |
C9H9BrN2O3 | |
DMSO |
Protect from light | |
White | |
188011-69-0 | |
273.1g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title