EMD Millipore Calbiochem™ Bisindolylmaleimide I, Hydrochloride

An enhanced water-soluble form of Bisindolylmaleimide I.

Manufacturer: EMD Millipore 203291250UG

 View more versions of this product

Catalog No. 20-329-1250UG

  Check Availability
Add to Cart

    Specifications

    Target Proteins Protein kinase C
    Molecular Formula C25H24N4O2·HCl
    Formula Weight 448.9g/mol
    Solubility DMSO or H2O
    Description 95% by HPLC
    Format Orange-red solid
    Quantity 250μg
    CAS 176504-36-2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit