EMD Millipore Calbiochem™ Bisindolylmaleimide V

A cell-permeable negative control compound for protein kinase C inhibition studies (IC50 >100 µM).

Manufacturer: EMD Millipore 203303250UG

 View more versions of this product

Catalog No. 20-330-3250UG

  Check Availability
Add to Cart

    Specifications

    Target Proteins Negative control compound for protein kinase C inhibition
    Molecular Formula C21H15N3O2
    Formula Weight 341.3g/mol
    Solubility DMSO or MeOH
    Description 95% by HPLC
    Format Redish orange solid
    Quantity 250μg
    CAS 113963-68-1

    Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit