EMD Millipore™ Calbiochem™ BQ-788, Sodium Salt

Highly selective ETB-receptor antagonist

Manufacturer: EMD Millipore™ 052238381MG

 View more versions of this product

Catalog No. 05-223-8381MG

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Quantity 1mg
    Product Type BQ-788, Sodium Salt
    Storage Requirements -20°C
    Format White to off-white lyophilized solid

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit