EMD Millipore Calbiochem™ Bradykinin

Induces the release of nitric oxide.

Manufacturer: EMD Millipore 052305005MG

 View more versions of this product

Catalog No. 05-230-5005MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Hygroscopic
    Formulation Supplied as an acetate salt.
    Description 97% by HPLC
    Quantity 5mg
    Target Proteins Induces the release of nitric oxide
    CAS 58-82-2
    Molecular Formula C50H73N15O11
    Formula Weight 1060.2g/mol
    Solubility 5% Acetic Acid
    Format White to off-white lyophilized solid

    Peptide Sequence: H-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit