Learn More
MilliporeSigma™ Calbiochem™ BRL 44408 Maleate
A selective α2A -adrenoceptor antagonist (Ki
Manufacturer:  MilliporeSigma™ 5.04795.0001
Catalog No. 50-479-50001
Add to cart
Description
Description
A selective α2A-adrenoceptor antagonist (Ki = 1.7nM and 144.5nM at α2A and α2B-adrenergic receptors respectively). Has been used to study regulation of neurotransmitter release from sympathetic nerves and shown to increase hippocampal noradrenaline release following systemic administration.
Specifications
Specifications
98% by HPLC
Hygroscopic
Protect from Light
Protect from Moisture
→2A-adrenoceptor
331.37g/mol
Tan powder
10mg
C13H17N3·C4H4O4
H2O