Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem c-Myc Inhibitor II
The c-Myc Inhibitor II, also referenced under CAS 413611-93-5, controls the biological activity of c-Myc.
Manufacturer: MilliporeSigma 47595710MG
Description
Packaged Under Inert GasSpecifications
95% by HPLC | |
Protect from Light | |
413611-93-5 | |
332.3g/mol |
Deep orange solid | |
10mg | |
C18H12N4O3 | |
DMSO or EtOH |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title