Learn More
MilliporeSigma™ Calbiochem™ c-Myc Inhibitor III, Mycro2
The c-Myc Inhibitor III, Mycro2 controls the biological activity of c-Myc.
Manufacturer:  MilliporeSigma™ 47596510MG
Catalog No. 47-596-510MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
99% by HPLC
Protect from Light
α-helical dimerization motifs of c-Myc/Max factors
408.4g/mol
Yellow solid
10mg
C17H11F3N4OS2
DMSO