Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Calmodulin Binding Domain
Potent calmodulin antagonist.
Supplier: MilliporeSigma™ 2087341MG
Description
Peptide Sequence: H-Leu-Lys-Lys-Phe-Asn-Ala-Arg-Arg-Lys-Leu-Lys-Gly-Ala-Ile-Leu-Thr-Thr-Met-Leu-Ala-OHSpecifications
Hygroscopic | |
Lyophilized Solid | |
Supplied as a trifluoroacetate salt. | |
Calmodulin dependent Protein Kinase-2 | |
C103H185N31O24S | |
H2O |
1 mg | |
White | |
95% by HPLC | |
115044-69-4 | |
2274.3g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title