EMD Millipore Calbiochem™ Calpain Inhibitor XI

The Calpain Inhibitor XI, also referenced under CAS 145731-49-3, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Manufacturer: EMD Millipore 2087431MG

 View more versions of this product

Catalog No. 20-874-31MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Hygroscopic
    Target Proteins calpain 1, calpain 2
    Molecular Formula C26H40N4O6
    Formula Weight 504.6g/mol
    Solubility DMSO
    Formulation Supplied as a trifluoroacetate salt.
    Description 95% by HPLC
    Format White to off-white solid
    Quantity 1mg
    CAS 145731-49-3

    • Packaged Under Inert Gas
    • Peptide Sequence: Z-Leu-Abu-CONH(CH2)3-morpholine (Abu = α-aminobutyric acid)
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit