Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Bradykinin
Induces the release of nitric oxide.
$28.50 - $220.00
Specifications
Color | White to Off-white |
---|---|
Formulation | Supplied as an acetate salt. |
Description | 97% by HPLC |
Target Proteins | Induces the release of nitric oxide |
CAS | 58-82-2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
05-230-5001MG
|
MilliporeSigma™
052305001MG |
1 mg |
Each for $28.50
|
|
|||||
05-230-5005MG
|
MilliporeSigma™
052305005MG |
5 mg |
Each for $88.00
|
|
|||||
05-230-50025M
|
MilliporeSigma™
0523050025MG |
25 mg |
Each for $220.00
|
|
|||||
Description
Peptide Sequence: H-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OHSpecifications
White to Off-white | |
97% by HPLC | |
58-82-2 | |
1060.2g/mol | |
Hygroscopic |
Supplied as an acetate salt. | |
Induces the release of nitric oxide | |
C50H73N15O11 | |
5% Acetic Acid | |
Lyophilized |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title