Learn More
MilliporeSigma™ Calbiochem™ cAMP Direct Immunoassay Kit (Colorimetric)
Competitive, Non-radioactive immunoassay kit used for rapid quantitative measurement of cAMP
Manufacturer:  MilliporeSigma™ 1168111KIT
Catalog No. 11-681-11KIT
Add to cart
Description
Description
  • Colorimetric method of detection
  • Released cAMP can be analyzed directly in the microtiter plate
  • Packaging: Fibre Case
Specifications
Specifications
cAMP Direct Immunoassay Kit (Colorimetric)
One Antibody Coated 96-Well Plate, cAMP Direct Conjugate, Anti-cAMP, cAMP Standard, 0.1N HCl, Neutralizing Reagent, Wash Buffer, Color Substrate, Stop Solution, Triethylamine, Acetic Anhydride, Plate Sealer, and user protocol
1 Kit/100 Tests
96-well format