Learn More
EMD Millipore Calbiochem™ Carboxyatractyloside, Potassium Salt, Xanthium sibiricum
Toxic compound isolated from the plant Xanthium sibiricum.
Manufacturer:  EMD Millipore 2162015MG
 View more versions of this product
Catalog No. 21-620-15MG
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
95% by HPLC
Hygroscopic
Protect from Moisture
35988-42-2
939.1g/mol
White solid
5mg
C31H43O18S2K3·3H2O
H2O