EMD Millipore Calbiochem™ Carboxyatractyloside, Potassium Salt, Xanthium sibiricum

Toxic compound isolated from the plant Xanthium sibiricum.

Manufacturer: EMD Millipore 2162012MG

 View more versions of this product

Catalog No. 21-620-12MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Hygroscopic
    Protect from Moisture
    Molecular Formula C31H43O18S2K3·3H2O
    Formula Weight 939.1g/mol
    Solubility H2O
    Description 95% by HPLC
    Format White solid
    Quantity 2mg
    CAS 35988-42-2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit