Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Casein Kinase I Inhibitor VIII
A cell-permeable, ATP site-targeting, γ isotype-selective CK1 inhibitor (IC50 = 5 nM).
Supplier: MilliporeSigma™ 5.00494.0001
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach
Specifications
Off-white | |
CKIγ | |
DMSO | |
Solid |
98% by HPLC | |
C20H17F2N3O3 | |
Protect from light | |
10 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title