Learn More
EMD Millipore Calbiochem™ Caspase-1 Inhibitor I
The Caspase-1 Inhibitor I, also referenced under CAS 143313-51-3, controls the biological activity of Caspase-1. This small molecule/inhibitor is primarily used for Cancer applications.
Manufacturer:  EMD Millipore 4000105MG
 View more versions of this product
Catalog No. 40-001-05MG
  Check Availability
Add to cart
Specifications
Description
Peptide Sequence: Ac-Tyr-Val-Ala-Asp-CHO
Description & Specifications
Specifications
97% by HPLC
Protect from Light
caspase-1, human recombinant
C23H32N4O8
DMSO or H2O
White powder
5mg
143313-51-3
492.5g/mol