EMD Millipore Calbiochem™ Caspase-1 Inhibitor I

The Caspase-1 Inhibitor I, also referenced under CAS 143313-51-3, controls the biological activity of Caspase-1. This small molecule/inhibitor is primarily used for Cancer applications.

Manufacturer: EMD Millipore 4000101MG

 View more versions of this product

Catalog No. 40-001-01MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins caspase-1, human recombinant
    Molecular Formula C23H32N4O8
    Formula Weight 492.5g/mol
    Solubility DMSO or H2O
    Description 97% by HPLC
    Format White powder
    Quantity 1mg
    CAS 143313-51-3

    Peptide Sequence: Ac-Tyr-Val-Ala-Asp-CHO

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit