EMD Millipore Calbiochem™ Caspase-3 Inhibitor IV

The Caspase-3 Inhibitor IV, also referenced under CAS 259199-63-8, controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.

Manufacturer: EMD Millipore 2354211MG

 View more versions of this product

Catalog No. 23-542-11MG

  Check Availability
Add to Cart

    Specifications

    Target Proteins caspase-3
    Molecular Formula C20H31N5O10S
    Formula Weight 533.6g/mol
    Solubility DMSO or H2O
    Description 95% by HPLC
    Format White lyophilized solid
    Quantity 1mg
    CAS 259199-63-8

    Peptide Sequence: Ac-Asp-Met-Gln-Asp-CHO

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit